Gene Transcripts

Gene Symbol
Tbc1d19
Entrez Gene ID
67249
Gene Name
TBC1 domain family, member 19
Gene Aliases
2810453K03Rik
Chromosome Location
Chr. 5: 53809581 - 53909221 on Build GRCm38
UniGene ID
Mm.28823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144517.4 NP_653100.1 18, 19 1455
GenBank mRNA AK012836.1 - 18, 19 1406
AK076128.1 - 18, 19 1403
AK083084.1 - 18, 19 1426
AK212437.1 - 1, 2 52
BC020138.1 - 18, 19 1414 3983701

View Full Product Details