Gene Transcripts
- Gene Symbol
- Agtpbp1
- Entrez Gene ID
- 67269
- Gene Name
- ATP/GTP binding protein 1
- Gene Aliases
- 1700020N17Rik, 2310001G17Rik, 2900054O13Rik, 4930445M19Rik, 5730402G09Rik, CCP1, Nna1, nmf243, pcd
- Chromosome Location
- Chr. 13: 59449534 - 59557364 on Build GRCm38
- UniGene ID
- Mm.153008
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048008.2 |
NP_001041473.1
|
13, 14 |
1470 |
|
NM_001284218.1 |
NP_001271147.1
|
13, 14 |
1595 |
|
NM_001284219.1 |
NP_001271148.1
|
12, 13 |
1541 |
|
NM_001284221.1 |
NP_001271150.1
|
12, 13 |
1345 |
|
NM_023328.3 |
NP_075817.2
|
13, 14 |
1470 |
|
GenBank mRNA |
AF219141.1 |
- |
13, 14 |
1409 |
|
AK051467.1 |
- |
12, 13 |
1381 |
|
AK132695.1 |
- |
13, 14 |
1410 |
|
AK133215.1 |
- |
13, 14 |
1529 |
|
AK170046.1 |
- |
13, 14 |
1413 |
|
BC042475.1 |
- |
12, 13 |
1541 |
|
BC060633.1 |
- |
13, 14 |
1514 |
|
BC082335.1 |
- |
12, 13 |
1333 |
30536788 |
CF729476.1 |
- |
4, 5 |
476 |
|
View Full Product Details