Gene Transcripts

Gene Symbol
Gpr89
Entrez Gene ID
67549
Gene Name
G protein-coupled receptor 89
Gene Aliases
4933412D19Rik, Gpr89a, SH120
Chromosome Location
Chr. 3: 96871066 - 96905343 on Build GRCm38
UniGene ID
Mm.46722
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026229.4 NP_080505.1 3 266
GenBank mRNA AK002937.1 - 3 286
AK009734.1 - 2 145
AK016787.1 - 3 266
BC010729.1 - 3 259 3708143

View Full Product Details