Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026334.3 | NP_080610.1 | 7 | 755 | |
GenBank mRNA | AK009300.1 | - | 7 | 757 | |
AK009359.1 | - | 7 | 757 | ||
AK009413.1 | - | 7 | 756 | ||
AK009428.1 | - | 7 | 759 | ||
AK009431.1 | - | 7 | 759 | ||
AK009437.1 | - | 7 | 757 | ||
AK009459.1 | - | 7 | 757 | ||
AK009473.1 | - | 7 | 756 | ||
AK009474.1 | - | 7 | 757 | ||
AK009479.1 | - | 7 | 754 | ||
AK009523.1 | - | 7 | 757 | ||
AK009525.1 | - | 7 | 757 | ||
AK009537.1 | - | 7 | 757 | ||
AK009546.1 | - | 7 | 760 | ||
AK009560.1 | - | 7 | 757 | ||
AK009571.1 | - | 7 | 756 | ||
AK009573.1 | - | 7 | 760 | ||
AK009729.1 | - | 7 | 757 | ||
AK009773.1 | - | 7 | 757 | ||
AK009875.1 | - | 7 | 757 | ||
AK009923.1 | - | 7 | 818 | ||
AK009990.1 | - | 7 | 757 | ||
AK010019.1 | - | 7 | 758 | ||
AK010026.1 | - | 7 | 757 | ||
AK010035.1 | - | 7 | 757 | ||
AK010036.1 | - | 7 | 756 | ||
AK010058.1 | - | 7 | 756 | ||
AK010061.1 | - | 7 | 757 | ||
AK010091.1 | - | 7 | 755 | ||
AK010093.1 | - | 7 | 757 | ||
AK010103.1 | - | 7 | 758 | ||
AK010106.1 | - | 7 | 757 | ||
AK010110.1 | - | 7 | 760 | ||
AK010116.1 | - | 7 | 755 | ||
AK010123.1 | - | 7 | 757 | ||
AK010124.1 | - | 6 | 751 | ||
AK010125.1 | - | 7 | 757 | ||
AK010139.1 | - | 7 | 757 | ||
AK010148.1 | - | 7 | 757 | ||
AK010173.1 | - | 7 | 757 | ||
AK010203.1 | - | 7 | 758 | ||
AK010231.1 | - | 7 | 756 | ||
AK010236.1 | - | 7 | 758 | ||
AK075913.1 | - | 7 | 757 | ||
BC061067.1 | - | 7 | 755 | 30301084 |