Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026836.2 | NP_081112.1 | 4 | 520 | |
GenBank mRNA | AK004168.1 | - | 3 | 167 | |
AK050689.1 | - | 4 | 521 | ||
AK133851.1 | - | 5 | 1504 | ||
AK183721.1 | - | 1 | 73 | ||
BC005603.1 | - | 4 | 499 | 3483315 |