Gene Transcripts

Gene Symbol
Arv1
Entrez Gene ID
68865
Gene Name
ARV1 homolog (yeast)
Gene Aliases
1110067L22Rik, AI461928, AW121084
Chromosome Location
Chr. 8: 124722138 - 124740648 on Build GRCm38
UniGene ID
Mm.22779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026855.4 NP_081131.1 2 243
GenBank mRNA AK004398.1 - 2 244
BC119376.1 - 2 223 8734125
BC120631.1 - 2 223 40129554
BC144875.1 - 2 223
BC144876.1 - 2 223

View Full Product Details