Gene Transcripts

Gene Symbol
Wdr53
Entrez Gene ID
68980
Gene Name
WD repeat domain 53
Gene Aliases
1500002B03Rik, AI848860
Chromosome Location
Chr. 16: 32247178 - 32257085 on Build GRCm38
UniGene ID
Mm.159860
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185162.1 NP_001172091.1 3 573
NM_026898.2 NP_081174.1 3 805
GenBank mRNA AK005105.1 - 3 772
BC028850.1 - 2 537 3987279

View Full Product Details