Gene Transcripts

Gene Symbol
Mtfmt
Entrez Gene ID
69606
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Gene Aliases
2310020P08Rik
Chromosome Location
Chr. 9: 65435562 - 65453054 on Build GRCm38
UniGene ID
Mm.287956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027134.3 NP_081410.2 3 526
GenBank mRNA AK009430.1 - 3 527
AK133492.1 - 3 521
BC019509.1 - 3 516 4456981

View Full Product Details