Gene Transcripts
- Gene Symbol
- Zc3h11a
- Entrez Gene ID
- 70579
- Gene Name
- zinc finger CCCH type containing 11A
- Gene Aliases
- 1110003F06Rik, 5730454B08Rik, G630041M05Rik, Zc3hdc11a, Zc3hh11a, mKIAA0663
- Chromosome Location
- Chr. 1: 133619868 - 133661399 on Build GRCm38
- UniGene ID
- Mm.245357
- Species
- Mus musculus
- Gene Symbol
- Zbed6
- Entrez Gene ID
- 667118
- Gene Name
- zinc finger, BED type containing 6
- Gene Aliases
- Gm8466, MGR
- Chromosome Location
- Chr. 1: 133655879 - 133660885 on Build GRCm38
- UniGene ID
- Mm.245357
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FR682664.1 |
- |
|
6189 |
|
View Full Product Details