Gene Transcripts

Gene Symbol
Cage1
Entrez Gene ID
71213
Gene Name
cancer antigen 1
Gene Aliases
4933427I01Rik, Ctag3
Chromosome Location
Chr. 13: 37994314 - 38036937 on Build GRCm38
UniGene ID
Mm.314688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027724.2 NP_082000.2 8, 9 2380
GenBank mRNA AK132733.1 - 4, 5 928
AY695827.1 - 7, 8 2115

View Full Product Details