Gene Transcripts

Gene Symbol
Chst9
Entrez Gene ID
71367
Gene Name
carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9
Gene Aliases
5430438D01Rik
Chromosome Location
Chr. 18: 15451773 - 15760415 on Build GRCm38
UniGene ID
Mm.329304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199055.2 NP_951010.1 4 1179
GenBank mRNA AB106879.1 - 4 1179
AK017407.1 - 6 1492
BC107361.1 - 4 1177 40056052

View Full Product Details