Gene Transcripts

Gene Symbol
R3hdm2
Entrez Gene ID
71750
Gene Name
R3H domain containing 2
Gene Aliases
1300003K24Rik, AU041262, mKIAA1002
Chromosome Location
Chr. 10: 127380332 - 127499384 on Build GRCm38
UniGene ID
Mm.29342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168292.1 NP_001161764.1 17 1851
NM_001168293.1 NP_001161765.1 15 2070
NM_027900.4 NP_082176.4 15 2031
GenBank mRNA AK004884.1 - 5 611
AK043545.1 - 15 2032
AK122416.1 - 16 1709
AX775759.1 - 6 728
BC043083.1 - 13 1409
BC053099.1 - 10 1114
BC064442.1 - 10 1082
BC082999.1 - 3 472
BC117927.2 - 15 1813 40097626

View Full Product Details