Gene Transcripts

Gene Symbol
Klhdc3
Entrez Gene ID
71765
Gene Name
kelch domain containing 3
Gene Aliases
1300011D16Rik, Peas
Chromosome Location
Chr. 17: 46674550 - 46680930 on Build GRCm38
UniGene ID
Mm.254784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163729.2 NP_001157201.2 3 534
NM_027910.3 NP_082186.2 4 687
GenBank mRNA AB053465.1 - 4 558
AJ278127.1 - 4 554
AK004964.1 - 4 664
AK079082.1 - 4 561
AK141673.1 - 3 519
AK170623.1 - 4 559
BC018154.1 - 4 656 3983436

View Full Product Details