Gene Transcripts

Gene Symbol
2310003H01Rik
Entrez Gene ID
71885
Gene Name
RIKEN cDNA 2310003H01 gene
Gene Aliases
Faap100
Chromosome Location
Chr. 11: 120369562 - 120378746 on Build GRCm38
UniGene ID
Mm.274890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027980.2 NP_082256.2 5 1612
GenBank mRNA AK082976.1 - 5 1613
AK147900.1 - 1701
AK148234.1 - 5 1597
AK165631.1 - 4 1483
BC034203.1 - 5 1483 4986832

View Full Product Details