Gene Transcripts

Gene Symbol
Tmub2
Entrez Gene ID
72053
Gene Name
transmembrane and ubiquitin-like domain containing 2
Gene Aliases
2010008E23Rik
Chromosome Location
Chr. 11: 102284820 - 102289427 on Build GRCm38
UniGene ID
Mm.101274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK008158.1 - 3 360
AK132118.1 - 3 405
BC016187.1 - 2 346
BC029841.1 - 2 663 4209499

View Full Product Details