Gene Transcripts

Gene Symbol
Cep89
Entrez Gene ID
72140
Gene Name
centrosomal protein 89
Gene Aliases
2610507L03Rik, Ccdc123
Chromosome Location
Chr. 7: 35397000 - 35439828 on Build GRCm38
UniGene ID
Mm.133263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028120.2 NP_082396.1 12, 13 1348
GenBank mRNA AK012067.1 - 12, 13 1349
AK029107.1 - 12, 13 1379
BC003981.1 - 12, 13 1403 3490246

View Full Product Details