Gene Transcripts

Gene Symbol
Trim29
Entrez Gene ID
72169
Gene Name
tripartite motif-containing 29
Gene Aliases
1110047J21Rik, 2810431N19Rik, 4732461M22Rik, AI119726
Chromosome Location
Chr. 9: 43310715 - 43336125 on Build GRCm38
UniGene ID
Mm.273277
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023655.2 NP_076144.2 2 948
GenBank mRNA AK028448.1 - 2 181
BB612224.1 - 2 178
BC006699.1 - 2 679
BC027353.1 - 2 863 3988926

View Full Product Details