Gene Transcripts

Gene Symbol
Ceacam18
Entrez Gene ID
72431
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 18
Gene Aliases
2010110O04Rik
Chromosome Location
Chr. 7: 43634670 - 43650228 on Build GRCm38
UniGene ID
Mm.158821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028236.1 NP_082512.1 2 356
GenBank mRNA AK008389.1 - 2 388
BC042780.1 - 2 356 4224563

View Full Product Details