Gene Transcripts
- Gene Symbol
- Ssbp3
- Entrez Gene ID
- 72475
- Gene Name
- single-stranded DNA binding protein 3
- Gene Aliases
- 2610021L12Rik, 2610200M23Rik, 5730488C10Rik, AI854733, AW551939, LAST, SSDP1
- Chromosome Location
- Chr. 4: 106911470 - 107049694 on Build GRCm38
- UniGene ID
- Mm.195635
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023672.2 |
NP_076161.2
|
11, 12 |
1140 |
|
NM_198438.1 |
NP_940840.1
|
10, 11 |
1059 |
|
GenBank mRNA |
AF170906.1 |
- |
10, 11 |
789 |
|
AF500117.1 |
- |
10, 11 |
789 |
|
AK011509.1 |
- |
11, 12 |
758 |
|
AK011853.1 |
- |
11, 12 |
1110 |
|
AK017711.1 |
- |
11, 12 |
1144 |
|
AK041388.1 |
- |
7, 8 |
656 |
|
AK133812.1 |
- |
8, 9 |
593 |
|
AK153733.1 |
- |
7, 8 |
500 |
|
AY167987.1 |
- |
10, 11 |
672 |
|
BC003430.1 |
- |
10, 11 |
813 |
2646149 |
BC028324.1 |
- |
5, 6 |
281 |
|
BC035475.1 |
- |
10, 11 |
1002 |
|
View Full Product Details