Gene Transcripts
- Gene Symbol
- Insig2
- Entrez Gene ID
- 72999
- Gene Name
- insulin induced gene 2
- Gene Aliases
- 2900053I11Rik, C730043J18Rik, Insig-2
- Chromosome Location
- Chr. 1: 121304353 - 121333654 on Build GRCm38
- UniGene ID
- Mm.27136
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271531.1 |
NP_001258460.1
|
5, 6 |
857 |
|
NM_001271532.1 |
NP_001258461.1
|
4, 5 |
724 |
|
NM_133748.2 |
NP_598509.1
|
5, 6 |
1100 |
|
NM_178082.3 |
NP_835183.1
|
5, 6 |
864 |
|
GenBank mRNA |
AF527631.1 |
- |
5, 6 |
834 |
|
AK040703.1 |
- |
5, 6 |
855 |
|
AK050394.1 |
- |
5, 6 |
858 |
|
AK152076.1 |
- |
5, 6 |
830 |
|
AK154543.1 |
- |
5, 6 |
863 |
|
AK161256.1 |
- |
5, 6 |
868 |
|
AK165634.1 |
- |
5, 6 |
869 |
|
AK191083.1 |
- |
3, 4 |
351 |
|
AK192853.1 |
- |
3, 4 |
309 |
|
AK203933.1 |
- |
3, 4 |
359 |
|
BC015288.1 |
- |
5, 6 |
814 |
4240711 |
BC023067.1 |
- |
5, 6 |
838 |
5357599 |
BC023874.1 |
- |
4, 5 |
756 |
5355039 |
BC024411.1 |
- |
5, 6 |
852 |
4971297 |
View Full Product Details