Gene Transcripts

Gene Symbol
Samd4
Entrez Gene ID
74480
Gene Name
sterile alpha motif domain containing 4
Gene Aliases
1700024G08Rik, 1700111L17Rik, 4933436G17Rik, Samd4a, Smaug, Smaug1
Chromosome Location
Chr. 14: 46882965 - 47105817 on Build GRCm38
UniGene ID
Mm.214454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037221.2 NP_001032298.1 3 1143
NM_001163433.1 NP_001156905.1 2 623
NM_028966.3 NP_083242.1 3 1143
GenBank mRNA AK034323.1 - 2 737
AK133470.1 - 3 1122
AK134243.1 - 3 1141
AK161277.1 - 2 617
AK163751.1 - 2 1164
BC145827.1 - 3 1009 40131130
DQ278487.1 - 3 920

View Full Product Details