Gene Transcripts

Gene Symbol
Mxra8
Entrez Gene ID
74761
Gene Name
matrix-remodelling associated 8
Gene Aliases
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam
Chromosome Location
Chr. 4: 155839680 - 155844102 on Build GRCm38
UniGene ID
Mm.159028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024263.4 NP_077225.4 4 573
GenBank mRNA AB040488.1 - 4 493
AB052097.1 - 4 451
AK004732.1 - 4 456
BC026438.1 - 4 495 4239626
EU849596.1 - 4 411
EU849597.1 - 4 402

View Full Product Details