Gene Transcripts

Gene Symbol
Nop14
Entrez Gene ID
75416
Gene Name
NOP14 nucleolar protein
Gene Aliases
2610033H07Rik, Nol14
Chromosome Location
Chr. 5: 34638536 - 34660148 on Build GRCm38
UniGene ID
Mm.40292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029278.2 NP_083554.2 17 2500
GenBank mRNA AK148605.1 - 17 2458
BC024998.1 - 17 2421 5012070
BC043043.1 - 17 2443 5701430

View Full Product Details