Gene Transcripts
- Gene Symbol
- Gns
- Entrez Gene ID
- 75612
- Gene Name
- glucosamine (N-acetyl)-6-sulfatase
- Gene Aliases
- 2610016K11Rik, AU042285, C87209, G6S, N28088
- Chromosome Location
- Chr. 10: 121365067 - 121397253 on Build GRCm38
- UniGene ID
- Mm.207683
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029364.3 |
NP_083640.1
|
13, 14 |
1667 |
|
GenBank mRNA |
AK030773.1 |
- |
13, 14 |
1666 |
|
AK049162.1 |
- |
13, 14 |
1668 |
|
AK054046.1 |
- |
13, 14 |
1667 |
|
AK083597.1 |
- |
13, 14 |
1645 |
|
AK151037.1 |
- |
11, 12 |
1289 |
|
AK159562.1 |
- |
13, 14 |
1634 |
|
AK165180.1 |
- |
13, 14 |
1647 |
|
AK165917.1 |
- |
13, 14 |
1617 |
|
AK169485.1 |
- |
13, 14 |
1649 |
|
AK170791.1 |
- |
13, 14 |
1663 |
|
AK192017.1 |
- |
2, 3 |
225 |
|
AK197879.1 |
- |
2, 3 |
225 |
|
BC055328.1 |
- |
13, 14 |
1661 |
6400020 |
BC059047.1 |
- |
5, 6 |
821 |
|
View Full Product Details