Gene Transcripts
- Gene Symbol
- Samd7
- Entrez Gene ID
- 75953
- Gene Name
- sterile alpha motif domain containing 7
- Gene Aliases
- 4930597A01Rik
- Chromosome Location
- Chr. 3: 30746293 - 30767174 on Build GRCm38
- UniGene ID
- Mm.308084
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029489.3 |
NP_083765.2
|
5, 6 |
1027 |
|
GenBank mRNA |
AK044227.1 |
- |
4, 5 |
903 |
|
View Full Product Details