Gene Transcripts

Gene Symbol
Slc35d3
Entrez Gene ID
76157
Gene Name
solute carrier family 35, member D3
Gene Aliases
6230421J19Rik, Frcl1
Chromosome Location
Chr. 10: 19847917 - 19852361 on Build GRCm38
UniGene ID
Mm.74324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029529.3 NP_083805.1 1 746
GenBank mRNA AK042761.1 - 1 498
AK044973.1 - 1 483
AK047610.1 - 1 747
BC139194.1 - 1 576 8862216

View Full Product Details