Gene Transcripts

Gene Symbol
Tgds
Entrez Gene ID
76355
Gene Name
TDP-glucose 4,6-dehydratase
Gene Aliases
2610017J16Rik, 2610025M23Rik, AI648925
Chromosome Location
Chr. 14: 118111911 - 118132765 on Build GRCm38
UniGene ID
Mm.251537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029578.3 NP_083854.3 12 1086
GenBank mRNA AK011439.1 - 11 974
AK154341.1 - 12 1063
AK165685.1 - 11 969
AK165955.1 - 12 1070
BC021419.1 - 12 1062 5012877

View Full Product Details