Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
76561
Gene Name
sorting nexin 7
Gene Aliases
2510028H01Rik
Chromosome Location
Chr. 3: 117781497 - 117868936 on Build GRCm38
UniGene ID
Mm.33824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190156.1 NP_001177085.1 3 398
NM_029655.3 NP_083931.2 3 398
GenBank mRNA AK011015.1 - 3 373
AK038971.1 - 3 399
AK052487.1 - 3 355
AK170966.1 - 3 345
BC087942.1 - 3 256 30303462

View Full Product Details