Gene Transcripts
- Gene Symbol
- Klhdc10
- Entrez Gene ID
- 76788
- Gene Name
- kelch domain containing 10
- Gene Aliases
- 2410127E18Rik, AI593221, mKIAA0265
- Chromosome Location
- Chr. 6: 30401517 - 30455179 on Build GRCm38
- UniGene ID
- Mm.277308
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029742.2 |
NP_084018.1
|
5, 6 |
855 |
|
GenBank mRNA |
AK010783.1 |
- |
5, 6 |
720 |
|
AK039732.1 |
- |
4, 5 |
757 |
|
AK050289.1 |
- |
3, 4 |
476 |
|
AK053834.1 |
- |
4, 5 |
727 |
|
AK085116.1 |
- |
4, 5 |
806 |
|
AK085130.1 |
- |
5, 6 |
813 |
|
AK172923.1 |
- |
4, 5 |
768 |
|
BC057064.1 |
- |
1, 2 |
806 |
|
BC060132.1 |
- |
5, 6 |
855 |
6409751 |
View Full Product Details