Gene Transcripts
- Gene Symbol
- Ninl
- Entrez Gene ID
- 78177
- Gene Name
- ninein-like
- Gene Aliases
- 4930519N13Rik, Gm1004, Gm1634, mKIAA0980
- Chromosome Location
- Chr. 2: 150934519 - 151009398 on Build GRCm38
- UniGene ID
- Mm.120911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207204.2 |
NP_997087.2
|
16 |
2211 |
|
GenBank mRNA |
BC067071.1 |
- |
16 |
2145 |
|
View Full Product Details