Gene Transcripts
- Gene Symbol
- Hhipl2
- Entrez Gene ID
- 78772
- Gene Name
- hedgehog interacting protein-like 2
- Gene Aliases
- 4930507C10Rik, AW208802, Hhip3
- Chromosome Location
- Chr. 1: 183413929 - 183437053 on Build GRCm38
- UniGene ID
- Mm.29003
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019671.1 |
- |
2 |
687 |
|
BC034362.1 |
- |
1 |
432 |
3154701 |
View Full Product Details