Gene Transcripts

Gene Symbol
Srd5a1
Entrez Gene ID
78925
Gene Name
steroid 5 alpha-reductase 1
Gene Aliases
0610031P22Rik, 4930435F02Rik, S5AR 1, Srd5a-1
Chromosome Location
Chr. 13: 69573449 - 69611900 on Build GRCm38
UniGene ID
Mm.451912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175283.3 NP_780492.2 1 228
GenBank mRNA AK082819.1 - 1 120
BC079863.1 - 1 206 6807475

View Full Product Details