Gene Transcripts

Gene Symbol
Pcdhb4
Entrez Gene ID
93875
Gene Name
protocadherin beta 4
Gene Aliases
Pcdhb5A, PcdhbD
Chromosome Location
Chr. 18: 37307455 - 37311173 on Build GRCm38
UniGene ID
Mm.196711
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053129.3 NP_444359.3 1 1477
GenBank mRNA AK029277.1 - 1 1469
AK036169.1 - 1 1478
AK143280.1 - 1 1478
AK160730.1 - 1 1478
AY013786.1 - 1 1293
BC053098.1 - 1 1450 6401988

View Full Product Details