Gene Transcripts

Gene Symbol
Pcdhb15
Entrez Gene ID
93886
Gene Name
protocadherin beta 15
Gene Aliases
Pcdhb7, PcdhbO
Chromosome Location
Chr. 18: 37473546 - 37476340 on Build GRCm38
UniGene ID
Mm.283857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053140.3 NP_444370.1 1 1175
GenBank mRNA AK038663.1 - 1 1177
AY013776.1 - 1 1004
BC066178.1 - 1 1181 30100073

View Full Product Details