Gene Transcripts

Gene Symbol
Cnnm4
Entrez Gene ID
94220
Gene Name
cyclin M4
Gene Aliases
5430430O18Rik, Acdp4
Chromosome Location
Chr. 1: 36471597 - 36508776 on Build GRCm38
UniGene ID
Mm.422638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033570.2 NP_291048.2 1 516
GenBank mRNA AF216963.1 - 1 128
AK173209.1 - 1 493
BC139799.1 - 1 117

View Full Product Details