Gene Transcripts

Gene Symbol
Tiparp
Entrez Gene ID
99929
Gene Name
TCDD-inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, AW558171, PARP7
Chromosome Location
Chr. 3: 65528447 - 65555518 on Build GRCm38
UniGene ID
Mm.246398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178892.5 NP_849223.2 2 754
GenBank mRNA AK028529.1 - 2 755
AK032453.1 - 2 738
AK150248.1 - 2 736
AK162094.1 - 2 760
AK163347.1 - 2 794
BC003919.1 - 1 14
BC068173.1 - 2 728 30616032

View Full Product Details