Gene Transcripts

Gene Symbol
Nhlrc1
Entrez Gene ID
105193
Gene Name
NHL repeat containing 1
Gene Aliases
AI505271, B230309E09Rik, EPM2B
Chromosome Location
Chr. 13: 47012557 - 47014850 on Build GRCm38
UniGene ID
Mm.25593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175340.4 NP_780549.1 1 1146
GenBank mRNA AK045746.1 - 1 1117
BC062644.1 - 1 1115
BC118951.1 - 1 1117 40093176
BC118952.1 - 1 1115 40093177

View Full Product Details