Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033541.4 | NP_291019.1 | 3, 4 | 822 | |
GenBank mRNA | AB067528.1 | - | 2, 3 | 479 | |
AF459815.1 | - | 2, 3 | 479 | ||
AF459816.1 | - | 2, 3 | 479 | ||
AK028986.1 | - | 3, 4 | 823 | ||
AK078952.1 | - | 2, 3 | 490 | ||
AK163253.1 | - | 2, 3 | 495 | ||
BC108955.1 | - | 2, 3 | 488 | 40046017 | |
BC126877.1 | - | 2, 3 | 487 | 40046021 |