Gene Transcripts

Gene Symbol
Acvrl1
Entrez Gene ID
11482
Gene Name
activin A receptor, type II-like 1
Gene Aliases
AI115505, AI427544, Acvrlk1, Alk1
Chromosome Location
Chr. 15: 101128522 - 101145336 on Build GRCm38
UniGene ID
Mm.279542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277255.1 NP_001264184.1 6 626
NM_001277257.1 NP_001264186.1 5 565
NM_001277258.1 NP_001264187.1 5 534
NM_001277259.1 NP_001264188.1 6 621
NM_009612.3 NP_033742.2 5 603
GenBank mRNA AK018754.1 - 2 105
AK134712.1 - 3 359
AK160915.1 - 6 594
AK170870.1 - 3 359
BC014291.1 - 4 522 3982004
BC015083.1 - 4 484 3980151
CT010330.1 - 3 331
L48015.1 - 5 605
Z31664.1 - 5 571

View Full Product Details