Gene Transcripts
- Gene Symbol
- Ang
- Entrez Gene ID
- 11727
- Gene Name
- angiogenin, ribonuclease, RNase A family, 5
- Gene Aliases
- AI385586, Ang1, Rnase5, Rnase5a
- Chromosome Location
- Chr. 14: 51091077 - 51102009 on Build GRCm38
- UniGene ID
- Mm.202665
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK171251.1 |
- |
2 |
535 |
|
BC055355.1 |
- |
2 |
491 |
5136641 |
View Full Product Details