Gene Transcripts

Gene Symbol
Ap3b1
Entrez Gene ID
11774
Gene Name
adaptor-related protein complex 3, beta 1 subunit
Gene Aliases
AP-3, AU015684, C78395, Hps2, beta3A, pe, pearl, rim2
Chromosome Location
Chr. 13: 94358952 - 94566316 on Build GRCm38
UniGene ID
Mm.21185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009680.3 NP_033810.2 23 2803
GenBank mRNA AF103809.2 - 23 2740
AK163996.1 - 8 861
BC015068.1 - 23 2809 4018563

View Full Product Details