Gene Transcripts
- Gene Symbol
- Cdk4
- Entrez Gene ID
- 12567
- Gene Name
- cyclin-dependent kinase 4
- Gene Aliases
- Crk3
- Chromosome Location
- Chr. 10: 127063537 - 127067288 on Build GRCm38
- UniGene ID
- Mm.6839
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009870.3 |
NP_034000.1
|
7, 8 |
993 |
|
GenBank mRNA |
AK003804.1 |
- |
7, 8 |
960 |
|
AK011871.1 |
- |
7, 8 |
981 |
|
AK012064.1 |
- |
7, 8 |
962 |
|
AK012202.1 |
- |
1, 2 |
79 |
|
AK019380.1 |
- |
7, 8 |
946 |
|
AK078440.1 |
- |
7, 8 |
947 |
|
AK146107.1 |
- |
7, 8 |
960 |
|
AK153465.1 |
- |
7, 8 |
993 |
|
AK218927.1 |
- |
3, 4 |
280 |
|
BC031599.1 |
- |
6, 7 |
1669 |
|
BC046336.1 |
- |
7, 8 |
993 |
6312184 |
BC052694.1 |
- |
7, 8 |
988 |
5714971 |
L01640.1 |
- |
6, 7 |
820 |
|
View Full Product Details