Gene Transcripts

Gene Symbol
Ctsg
Entrez Gene ID
13035
Gene Name
cathepsin G
Gene Aliases
Chromosome Location
Chr. 14: 56099881 - 56102574 on Build GRCm38
UniGene ID
Mm.4858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007800.1 NP_031826.1 2 253
GenBank mRNA BC125511.1 - 2 168
BC125513.1 - 2 168 40130028
X78544.1 - 2 253

View Full Product Details