Gene Transcripts

Gene Symbol
Cyp2b19
Entrez Gene ID
13090
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 19
Gene Aliases
CYPIIB19
Chromosome Location
Chr. 7: 26757187 - 26772630 on Build GRCm38
UniGene ID
Mm.14098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF047529.1 - 1 12
AK029227.1 - 1 37
BC137965.1 - 1 57 8860984
BC137966.1 - 1 57 8860985

View Full Product Details