Gene Transcripts

Gene Symbol
Cyp2c29
Entrez Gene ID
13095
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 29
Gene Aliases
AHOH, AHOHase, AI159681, AI196010, Ah-2, Ahh-1, Cyp2c, P450-2C
Chromosome Location
Chr. 19: 39287044 - 39330713 on Build GRCm38
UniGene ID
Mm.20764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007815.3 NP_031841.3 5 822
GenBank mRNA AK139760.1 - 862
AK149558.1 - 5 641
BC019908.1 - 5 804 5123631
D17674.1 - 5 796

View Full Product Details