Gene Transcripts
- Gene Symbol
- Cyp3a16
- Entrez Gene ID
- 13114
- Gene Name
- cytochrome P450, family 3, subfamily a, polypeptide 16
- Gene Aliases
- Chromosome Location
- Chr. 5: 145436309 - 145469737 on Build GRCm38
- UniGene ID
- Mm.378905
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007820.2 |
NP_031846.2
|
5 |
435 |
|
GenBank mRNA |
D26137.1 |
- |
5 |
406 |
|
View Full Product Details