Gene Transcripts

Gene Symbol
Fkbp2
Entrez Gene ID
14227
Gene Name
FK506 binding protein 2
Gene Aliases
13kDa, FKBP-2, mFKBP13, mFKBP2
Chromosome Location
Chr. 19: 6977739 - 6980501 on Build GRCm38
UniGene ID
Mm.4234
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166368.1 NP_001159840.1 2 207
NM_008020.3 NP_032046.1 2 222
GenBank mRNA AK144284.1 - 2 223
AK165394.1 - 2 187
BC031824.1 - 2 176 4196363
BC053692.1 - 2 212 6395841
CF949699.1 - 1 26
DV655383.1 - 2 207

View Full Product Details