Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159551.1 | NP_001153023.1 | 3 | 436 | |
NM_001159552.1 | NP_001153024.1 | 3 | 436 | ||
NM_001159553.1 | NP_001153025.1 | 3 | 436 | ||
NM_010376.4 | NP_034506.1 | 3 | 436 | ||
GenBank mRNA | AF483214.1 | - | 3 | 403 | |
AF515662.1 | - | 3 | 315 | ||
AJ345032.1 | - | 3 | 446 | ||
AK004690.1 | - | 3 | 410 | ||
AK007664.1 | - | 3 | 418 | ||
AK012600.1 | - | 3 | 429 | ||
AK019877.1 | - | 3 | 410 | ||
AK028109.1 | - | 3 | 405 | ||
AK039008.1 | - | 3 | 410 | ||
AK049101.1 | - | 3 | 410 | ||
AK050664.1 | - | 3 | 379 | ||
AK051151.1 | - | 3 | 388 | ||
AK080966.1 | - | 3 | 398 | ||
AK159176.1 | - | 3 | 410 | ||
AK169922.1 | - | 3 | 393 | ||
AK171790.1 | - | 3 | 364 | ||
BC056977.1 | - | 3 | 435 | 6818912 | |
CA321999.1 | - | 3 | 436 | ||
DQ168449.1 | - | 3 | 452 |