Gene Transcripts

Gene Symbol
Hoxc5
Entrez Gene ID
15424
Gene Name
homeobox C5
Gene Aliases
AI181837, Hox-3.4, Hox-6.2
Chromosome Location
Chr. 15: 103014008 - 103017429 on Build GRCm38
UniGene ID
Mm.103825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175730.5 NP_783857.1 1 206
GenBank mRNA AK078527.1 - 1 207
BC115551.1 - 1 207 40101712
BC115552.1 - 1 207 40101713

View Full Product Details