Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253872.1 | NP_001240801.1 | 13 | 1598 | |
NM_001253873.1 | NP_001240802.1 | 13 | 1598 | ||
NM_001253874.1 | NP_001240803.1 | 13 | 1598 | ||
NM_008400.3 | NP_032426.2 | 13 | 1598 | ||
GenBank mRNA | AF065901.1 | - | 13 | 1400 | |
AF065902.1 | - | 13 | 1400 | ||
AK141575.1 | - | 3 | 289 | ||
AK152913.1 | - | 13 | 1398 | ||
AK154115.1 | - | 13 | 1533 | ||
AK155566.1 | - | 13 | 1482 | ||
AK156251.1 | - | 13 | 1568 | ||
AK169628.1 | - | 13 | 1514 | ||
AK171389.1 | - | 13 | 1482 | ||
AK172394.1 | - | 13 | 1544 | ||
BC145094.1 | - | 13 | 1412 | 9053642 | |
BC145095.1 | - | 13 | 1412 | 9053643 | |
BC145096.1 | - | 13 | 1412 | 9053644 | |
BC145802.1 | - | 13 | 1412 | 40131105 | |
JN986841.1 | - | 13 | 1393 | ||
M60778.1 | - | 13 | 1448 |